1,4-Dibutylbenzene ≥98.0%

Supplier: TCI America

D2781-25ML D2781-5ML
TCD2781-25MLEA 736.28 CAD
TCD2781-25ML TCD2781-5ML
1,4-Dibutylbenzene ≥98.0%
1,4-Dibutylbenzene

CAS Number: 1571-86-4
MDL Number: MFCD01076591
Molecular Formula: C14H22
Molecular Weight: 190.33
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.86

Formula: C₁₄H₂₂
MW: 190.33 g/mol
Density: 0.86 (20 °C)
MDL Number: MFCD01076591
CAS Number: 1571-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e0a48c3eea08ece044ca1d8e1f8eb73cf8423c&vid=a0e0a48c3eea08ece044ca1d8e1f8eb73cf8423c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR