3,5-Dibenzyloxybenzyl alcohol ≥98.0%

Supplier: TCI America

D2651-25G D2651-5G
TCD2651-25GEA 406.06 CAD
TCD2651-25G TCD2651-005G
3,5-Dibenzyloxybenzyl alcohol ≥98.0%
3,5-Dibenzyloxybenzyl alcohol

CAS Number: 24131-31-5
MDL Number: MFCD01863236
Molecular Formula: C21H20O3
Molecular Weight: 320.39
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: 3,5-Bis(benzyloxy)benzyl Alcohol

Formula: C₂₁H₂₀O₃
Melting Pt: 82 °C
MDL Number: MFCD01863236
CAS Number: 24131-31-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51c83db5db9d6e2e26228e58a9b0c6e81d392833&vid=51c83db5db9d6e2e26228e58a9b0c6e81d392833&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR