4,5-Dimethoxy-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2628-5G D2628-25G
TCD2628-005GEA 84.37 CAD
TCD2628-005G TCD2628-25G
4,5-Dimethoxy-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
4,5-Dimethoxy-2-nitrobenzoic acid

CAS Number: 4998-07-6
MDL Number: MFCD00014697
Molecular Formula: C9H9NO6
Molecular Weight: 227.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 194

Formula: C₉H₉NO₆
Melting Pt: 194 °C
MDL Number: MFCD00014697
CAS Number: 4998-07-6
EINECS: 225-657-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f87fb345683e723e21cbea7baae65a67cac3f5a&vid=3f87fb345683e723e21cbea7baae65a67cac3f5a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR