Dimethylmalonic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D2606-5G
D2606-25G
TCD2606-005GEA
39.5
CAD
TCD2606-005G
TCD2606-025G
Dimethylmalonic acid ≥98.0% (by GC, titration analysis)
Dimethylmalonic acid
CAS Number: 595-46-0
MDL Number: MFCD00004193
Molecular Formula: C5H8O4
Molecular Weight: 132.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 191
Formula:
C₅H₈O₄ Melting Pt: 191 °C |
MDL Number:
MFCD00004193 CAS Number: 595-46-0 EINECS: 209-867-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...