3,5-Di-tert-butylaniline ≥98.0%
Supplier: TCI America
|
Warning
|
D2515-5G
D2515-1G
TCD2515-5GEA
630.42
CAD
TCD2515-5G
TCD2515-001G
3,5-Di-tert-butylaniline ≥98.0%
3,5-Di-tert-butylaniline
CAS Number: 2380-36-1
MDL Number: MFCD00134096
Molecular Formula: C14H23N
Molecular Weight: 205.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 55
Flash Point (°C): 110
Formula:
C₁₄H₂₃N Melting Pt: 55 °C Flash Pt: 110 °C |
MDL Number:
MFCD00134096 CAS Number: 2380-36-1 EINECS: 219-173-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...