1,11-Dibromoundecane ≥96.0%

Supplier: TCI America

Synonyms: Undecamethylene dibromide

D2487-1G D2487-5G
TCD2487-1GEA 39.5 CAD
TCD2487-1G TCD2487-005G
1,11-Dibromoundecane ≥96.0%
1,11-Dibromoundecane

CAS Number: 16696-65-4
MDL Number: MFCD00000224
Molecular Formula: C11H22Br2
Molecular Weight: 314.11
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 192
Melting point (°C): -11
Flash Point (°C): 110
Specific Gravity (20/20): 1.34


Other Product Names / Synonyms: Undecamethylene Dibromide

Formula: BrH₂C(CH₂)₉CH₂Br
MW: 314.1 g/mol
Boiling Pt: 192 °C
Melting Pt: –11 °C
Density: 1.34 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000224
CAS Number: 16696-65-4
EINECS: 240-745-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b24f5653ad29dd461dd7af2eb7aacb74b6537c62&vid=b69e5a5bbf44f4875a7d5571263b6dab58f30ea4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR