2,6-Di-tert-butylnaphthalene ≥98.0%
Supplier: TCI America
D2369-5G
D2369-25G
TCD2369-5GEA
153.26
CAD
TCD2369-5G
TCD2369-025G
2,6-Di-tert-butylnaphthalene ≥98.0%
2,6-Di-tert-butylnaphthalene
CAS Number: 3905-64-4
MDL Number: MFCD00004110
Molecular Formula: C18H24
Molecular Weight: 240.39
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 148
Formula:
C₁₈H₂₄ Melting Pt: 148 °C |
MDL Number:
MFCD00004110 CAS Number: 3905-64-4 EINECS: 223-461-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...