Diethyl phosphoramidate ≥98.0%
Supplier: TCI America
D2361-25G
D2361-5G
TCD2361-025GEA
207.31
CAD
TCD2361-025G
TCD2361-5G
Diethyl phosphoramidate ≥98.0%
Diethyl phosphoramidate
CAS Number: 1068-21-9
MDL Number: MFCD00015676
Molecular Formula: C4H12NO3P
Molecular Weight: 153.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 52
Flash Point (°C): 110
Product Categories: Synthetic Organic Chemistry, Amination
Other Product Names / Synonyms: Diethyl Amidophosphate, Diethyl Phosphoryl Amide
Formula:
C₄H₁₂NO₃P Boiling Pt: 140 °C Melting Pt: 52 °C Flash Pt: 110 °C |
MDL Number:
MFCD00015676 CAS Number: 1068-21-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...