3',4'-Dihydroxyacetophenone ≥98.0%

Supplier: TCI America

Synonyms: 1-(3,4-Dihydroxyphenyl)ethan-1-one, Acetopyrocatechol

D2345-25G D2345-5G
TCD2345-25GEA 567.22 CAD
TCD2345-25G TCD2345-005G
3',4'-Dihydroxyacetophenone ≥98.0%
3',4'-Dihydroxyacetophenone

CAS Number: 1197-09-7
MDL Number: MFCD00016439
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 119


Other Product Names / Synonyms: 4-Acetocatechol, 4-Acetylpyrocatechol

Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 119 °C
MDL Number: MFCD00016439
CAS Number: 1197-09-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d9cf6a6280e72e7e4bcc2a06b7fc5f44937b700&vid=7d9cf6a6280e72e7e4bcc2a06b7fc5f44937b700&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR