1,2-Dihydroxynaphthalene ≥95.0%
Supplier: TCI America
|
Warning
|
D2299-1G
D2299-5G
TCD2299-1GEA
360.4
CAD
TCD2299-1G
TCD2299-005G
1,2-Dihydroxynaphthalene ≥95.0%
1,2-Dihydroxynaphthalene
CAS Number: 574-00-5
MDL Number: MFCD00003959
Molecular Formula: C10H8O2
Molecular Weight: 160.17
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 103
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 1,2-Naphthalenediol
Formula:
C₁₀H₈O₂ Melting Pt: 103 °C |
MDL Number:
MFCD00003959 CAS Number: 574-00-5 EINECS: 209-365-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...