Dicyclohexyl ketone ≥97.0%
Supplier: TCI America
D2258-500ML
D2258-25ML
TCD2258-500MLEA
231.47
CAD
TCD2258-500ML
TCD2258-025ML
Dicyclohexyl ketone ≥97.0%
Dicyclohexyl ketone
CAS Number: 119-60-8
MDL Number: MFCD00040417
Molecular Formula: C13H22O
Molecular Weight: 194.32
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 113
Specific Gravity (20/20): 0.96
Other Product Names / Synonyms: Dodecahydrobenzophenone
Formula:
C₁₃H₂₂O Boiling Pt: 135 °C Density: 0.96 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00040417 CAS Number: 119-60-8 EINECS: 204-336-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...