trans-1,2-Dichlorocyclohexane ≥96.0%
Supplier: TCI America
D2230-5G
D2230-25G
TCD2230-5GEA
210.2
CAD
TCD2230-5G
TCD2230-025G
trans-1,2-Dichlorocyclohexane ≥96.0%
trans-1,2-Dichlorocyclohexane
CAS Number: 822-86-6
MDL Number: MFCD00003824
Molecular Formula: C6H10Cl2
Molecular Weight: 153.05
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 69
Flash Point (°C): 66
Specific Gravity (20/20): 1.18
Formula:
C₆H₁₀Cl₂ Boiling Pt: 69 °C Melting Pt: –6 °C Density: 1.18 (20 °C) Flash Pt: 66 °C |
MDL Number:
MFCD00003824 CAS Number: 822-86-6 EINECS: 212-503-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...