2,6-Dichloro-p-cresol ≥96.0%
Supplier: TCI America
Synonyms:
2,6-Dichloro-4-methylphenol
D2173-5G
D2173-1G
TCD2173-005GEA
65.89
CAD
TCD2173-005G
TCD2173-1G
2,6-Dichloro-p-cresol ≥96.0%
2,6-Dichloro-p-cresol
CAS Number: 2432-12-4
MDL Number: MFCD00134679
Molecular Formula: C7H6Cl2O
Molecular Weight: 177.02
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Flash Point (°C): 120
Freezing point (°C): 34
Other Product Names / Synonyms: 2,6-Dichloro-4-methylphenol
Formula:
C₇H₆Cl₂O MW: 177.03 g/mol Flash Pt: 120 °C |
MDL Number:
MFCD00134679 CAS Number: 2432-12-4 EINECS: 219-402-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...