trans-2,3-Dibromo-2-butene-1,4-diol ≥99.0%

Supplier: TCI America

D2169-25G D2169-500G
TCD2169-025GEA 47.4 CAD
TCD2169-025G TCD2169-500G
trans-2,3-Dibromo-2-butene-1,4-diol ≥99.0%
trans-2,3-Dibromo-2-butene-1,4-diol

CAS Number: 21285-46-1
MDL Number: MFCD00004698
Molecular Formula: C4H6Br2O2
Molecular Weight: 245.90
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 114


Other Product Names / Synonyms: trans-2,3-Dibromo-1,4-dihydroxy-2-butene

Formula: C₄H₆Br₂O₂
MW: 245.9 g/mol
Melting Pt: 114 °C
MDL Number: MFCD00004698
CAS Number: 21285-46-1
EINECS: 244-313-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20ed3f295eea494beb092ecc6c0c1c489883060c&vid=20ed3f295eea494beb092ecc6c0c1c489883060c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR