1,4-Diethynylbenzene ≥98.0%

Supplier: TCI America

D2151-5G D2151-1G
TCD2151-005GEA 350.76 CAD
TCD2151-005G TCD2151-001G
1,4-Diethynylbenzene ≥98.0%
1,4-Diethynylbenzene

CAS Number: 935-14-8
MDL Number: MFCD00078375
Molecular Formula: C10H6
Molecular Weight: 126.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 92

Formula: C₁₀H₆
Melting Pt: 92 °C
MDL Number: MFCD00078375
CAS Number: 935-14-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96d8b3abebde65f72d36c7cb553b74d3353544ee&vid=96d8b3abebde65f72d36c7cb553b74d3353544ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR