2,4-Diaminopyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D2120-5G D2120-25G
TCD2120-005GEA 310.79 CAD
TCD2120-005G TCD2120-025G
2,4-Diaminopyrimidine ≥98.0% (by GC, titration analysis)
2,4-Diaminopyrimidine

CAS Number: 156-81-0
MDL Number: MFCD00038023
Molecular Formula: C4H6N4
Molecular Weight: 110.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 149


Other Product Names / Synonyms: 2,4-Pyrimidinediamine

Formula: C₄H₆N₄
Melting Pt: 149 °C
MDL Number: MFCD00038023
CAS Number: 156-81-0
EINECS: 205-862-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ff64c54cc2ee64dfc7769e991195cac38558df1&vid=6ff64c54cc2ee64dfc7769e991195cac38558df1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR