2,2-Diphenylethylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1-Amino-2,2-diphenylethane, 2,2-Diphenylethanamine
D2018-25G
D2018-5G
TCD2018-25GEA
420.28
CAD
TCD2018-25G
TCD2018-5G
2,2-Diphenylethylamine ≥98.0% (by GC, titration analysis)
2,2-Diphenylethylamine
CAS Number: 3963-62-0
MDL Number: MFCD00008143
Molecular Formula: C14H15N
Molecular Weight: 197.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 49
Flash Point (°C): 110
Other Product Names / Synonyms: 1-Amino-2,2-diphenylethane
Formula:
C₁₄H₁₅N MW: 197.28 g/mol Melting Pt: 49 °C Flash Pt: 110 °C |
MDL Number:
MFCD00008143 CAS Number: 3963-62-0 EINECS: 223-565-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...