3-Dodecylthiophene ≥98.0%

Supplier: TCI America

D2016-25G D2016-5G D2016-1G
TCD2016-25GEA 597.24 CAD
TCD2016-25G TCD2016-005G TCD2016-001G
3-Dodecylthiophene ≥98.0%
3-Dodecylthiophene

CAS Number: 104934-52-3
MDL Number: MFCD00130121
Molecular Formula: C16H28S
Molecular Weight: 252.46
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 113
Specific Gravity (20/20): 0.91


Product Categories: Materials Chemistry, Building Blocks for Semiconducting Polymers, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Laurylthiophene

Formula: C₁₆H₂₈S
Boiling Pt: 175 °C
Density: 0.91 (20 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130121
CAS Number: 104934-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af663f3aff9ff891c2d72f425260609dfaf6470f&vid=af663f3aff9ff891c2d72f425260609dfaf6470f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR