2,4-Dimethoxyaniline ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1982-25G D1982-100G
TCD1982-025GEA 43.56 CAD
TCD1982-025G TCD1982-100G
2,4-Dimethoxyaniline ≥98.0% (by titrimetric analysis)
2,4-Dimethoxyaniline

CAS Number: 2735-04-8
MDL Number: MFCD00008371
Molecular Formula: C8H11NO2
Molecular Weight: 153.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 33

Formula: C₈H₁₁NO₂
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 39 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008371
CAS Number: 2735-04-8
EINECS: 220-355-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e852705d13f07c36b9ea4f22a0b6de6a55361e3&vid=91b52744fa6856fc84aaf602c523da0d3b0e438b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR