1,6-Dibromo-2-naphthol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,6-Dibromo-2-hydroxynaphthalene, 1,6-Dibromonaphthalen-2-ol

D1872-250G D1872-25G
TCD1872-250GEA 932.2 CAD
TCD1872-250G TCD1872-025G
1,6-Dibromo-2-naphthol ≥97.0% (by GC, titration analysis)
1,6-Dibromo-2-naphthol

CAS Number: 16239-18-2
MDL Number: MFCD00003870
Molecular Formula: C10H6Br2O
Molecular Weight: 301.97
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 108

Formula: C₁₀H₆Br₂O
MW: 301.97 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 108 °C
MDL Number: MFCD00003870
CAS Number: 16239-18-2
EINECS: 240-356-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb48ed743d5eb0fd55a6fa3f9231ba891c324e13&vid=cb48ed743d5eb0fd55a6fa3f9231ba891c324e13&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR