2,4-Difluorobenzonitrile ≥98.0%

Supplier: TCI America

D1826-5G D1826-25G
TCD1826-005GEA 47.87 CAD
TCD1826-005G TCD1826-025G
2,4-Difluorobenzonitrile ≥98.0%
2,4-Difluorobenzonitrile

CAS Number: 3939-09-1
MDL Number: MFCD00009826
Molecular Formula: C7H3F2N
Molecular Weight: 139.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 83
Melting point (°C): 47
Flash Point (°C): 110

Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 83 °C
Melting Pt: 47 °C
Flash Pt: 110 °C
MDL Number: MFCD00009826
CAS Number: 3939-09-1
EINECS: 223-523-8
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d78371ac7b3817e2f69351d5eb11b887088d60fb&vid=60c29bc577c8b7a7701f1516479668a394f67a01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR