3,4-Difluoroaniline ≥98.0%

Supplier: TCI America
Warning

D1809-25G D1809-100G
TCD1809-025GEA 146.94 CAD
TCD1809-025G TCD1809-100G
3,4-Difluoroaniline ≥98.0%
3,4-Difluoroaniline

CAS Number: 3863-11-4
MDL Number: MFCD00007761
Molecular Formula: C6H5F2N
Molecular Weight: 129.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 85
Freezing point (°C): 22

Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 205
Melting Pt: 22
Density: 1.302 g/cm³ (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007761
CAS Number: 3863-11-4
EINECS: 223-381-7
UN: 2941
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb90f62a1ac8563dd2d0f19826f246d58ae098f8&vid=cb90f62a1ac8563dd2d0f19826f246d58ae098f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR