2,6-Di-tert-butylpyridine ≥97.0%

Supplier: TCI America
Warning

D1804-25G D1804-5G
TCD1804-25GEA 774.2 CAD
TCD1804-25G TCD1804-5G
2,6-Di-tert-butylpyridine ≥97.0%
2,6-Di-tert-butylpyridine

CAS Number: 585-48-8
MDL Number: MFCD00006306
Molecular Formula: C13H21N
Molecular Weight: 191.32
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 101
Melting point (°C): 2
Flash Point (°C): 71
Specific Gravity (20/20): 0.87

Formula: C₁₃H₂₁N
Boiling Pt: 101 °C
Melting Pt: 2 °C
Density: 0.87 (20 °C)
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006306
CAS Number: 585-48-8
EINECS: 209-557-6
Merck Index: 12,03086
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=862672e615c23030ce90fdfde5bceb0c4fa029d6&vid=862672e615c23030ce90fdfde5bceb0c4fa029d6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR