2,2'-Dimethylbiphenyl ≥95.0%

Supplier: TCI America

Synonyms: 2,2'-Bitolyl, 2,2'-Dimethyl-1,1'-biphenyl

D1717-5G D1717-1G
TCD1717-005GEA 208.56 CAD
TCD1717-005G TCD1717-001G
2,2'-Dimethylbiphenyl ≥95.0%
2,2'-Dimethylbiphenyl

CAS Number: 605-39-0
MDL Number: MFCD00048075
Molecular Formula: C14H14
Molecular Weight: 182.27
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 259
Freezing point (°C): 18
Specific Gravity (20/20): 0.99


Product Categories: Materials Chemistry, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: 2,2'-Bitolyl

Formula: C₁₄H₁₄
MW: 182.27 g/mol
Boiling Pt: 259 °C
Melting Pt: 18
Density: 0.99 (20 °C)
MDL Number: MFCD00048075
CAS Number: 605-39-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53191249a92547b0a84bfb091b64bae6720ac8b0&vid=53191249a92547b0a84bfb091b64bae6720ac8b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR