4,4'-Diaminobenzophenone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Bis(4-aminophenyl)methanone, 4,4′-Diaminobenzophenone

D1561-1G D1561-5G
TCD1561-001GEA 110.6 CAD
TCD1561-001G TCD1561-5G
4,4'-Diaminobenzophenone ≥98.0% (by HPLC, titration analysis)
4,4'-Diaminobenzophenone

CAS Number: 611-98-3
MDL Number: MFCD00038138
Molecular Formula: C13H12N2O
Molecular Weight: 212.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 245

Formula: C₁₃H₁₂N₂O
MW: 212.25 g/mol
Melting Pt: 245 °C
MDL Number: MFCD00038138
CAS Number: 611-98-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dddab5f9ee451d6e0602ee638e78cca932ba3411&vid=dddab5f9ee451d6e0602ee638e78cca932ba3411&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR