7-Tridecanone ≥97.0%
Supplier: TCI America
D1546-5G
D1546-25G
TCD1546-5GEA
94.8
CAD
TCD1546-5G
TCD1546-025G
7-Tridecanone ≥97.0%
7-Tridecanone
CAS Number: 462-18-0
MDL Number: MFCD00009543
Molecular Formula: C13H26O
Molecular Weight: 198.35
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 264
Flash Point (°C): 110
Freezing point (°C): 30
Other Product Names / Synonyms: Dihexyl Ketone, Enanthone
Formula:
C₁₃H₂₆O Boiling Pt: 264 °C Melting Pt: 30…33 °C Density: 0.825 g/cm³ (25 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009543 CAS Number: 462-18-0 EINECS: 207-324-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...