8-Pentadecanone ≥99.0%

Supplier: TCI America

Synonyms: Diheptyl ketone, Caprylone

D1545-5G D1545-25G
TCD1545-5GEA 191.18 CAD
TCD1545-5G TCD1545-025G
8-Pentadecanone ≥99.0%
8-Pentadecanone

CAS Number: 818-23-5
MDL Number: MFCD00009555
Molecular Formula: C15H30O
Molecular Weight: 226.40
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 43
Flash Point (°C): 110


Other Product Names / Synonyms: Caprylone, Diheptyl Ketone

Formula: C₁₅H₃₀O
MW: 226.4 g/mol
Boiling Pt: 140 °C
Melting Pt: 43 °C
Density: 0.787 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009555
CAS Number: 818-23-5
EINECS: 212-450-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea2ba9e2520d049e869a80999c107f368c7fcfc4&vid=699075ec0924be436a2476b40c3ad6d2a8bdc8d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR