Methyl-2,6-dihydroxybenzoate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D1461-25G
D1461-5G
TCD1461-25GEA
464.52
CAD
TCD1461-25G
TCD1461-005G
Methyl-2,6-dihydroxybenzoate ≥98.0% (by GC, titration analysis)
Methyl 2,6-dihydroxybenzoate
CAS Number: 2150-45-0
MDL Number: MFCD00059619
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 70
Flash Point (°C): 110
Other Product Names / Synonyms: 2,6-Dihydroxybenzoic Acid Methyl Ester, gamma-Resorcylic Acid Methyl Ester
Formula:
C₈H₈O₄ Melting Pt: 70 °C Flash Pt: 110 °C |
MDL Number:
MFCD00059619 CAS Number: 2150-45-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...