3,7-Dimethyl-1-octanol ≥98.0%
Supplier: TCI America
D1442-25ML
D1442-500ML
TCD1442-025MLEA
56.55
CAD
TCD1442-025ML
TCD1442-500ML
3,7-Dimethyl-1-octanol ≥98.0%
3,7-Dimethyl-1-octanol
CAS Number: 106-21-8
MDL Number: MFCD00002936
Molecular Formula: C10H22O
Molecular Weight: 158.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 110
Specific Gravity (20/20): 0.83
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Acyclic Monoterpenes
Other Product Names / Synonyms: Tetrahydrogeraniol
Formula:
C₁₀H₂₂O Boiling Pt: 118 °C Density: 0.83 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00002936 CAS Number: 106-21-8 EINECS: 203-374-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...