3,3'-Dimethylbiphenyl ≥98.0%
Supplier: TCI America
Synonyms:
3,3'-Bitolyl, 3,3'-Dimethyl-1,1'-biphenyl
D1430-5ML
D1430-25ML
TCD1430-005MLEA
233.05
CAD
TCD1430-005ML
TCD1430-25ML
3,3'-Dimethylbiphenyl ≥98.0%
3,3'-Dimethylbiphenyl
CAS Number: 612-75-9
MDL Number: MFCD00008534
Molecular Formula: C14H14
Molecular Weight: 182.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 110
Freezing point (°C): 9
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: 3,3'-Bitolyl
Formula:
C₁₄H₁₄ MW: 182.27 g/mol Boiling Pt: 155 °C Density: 1.00 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00008534 CAS Number: 612-75-9 EINECS: 210-319-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...