2,7-Dinitro-9-fluorenone ≥98.0%

Supplier: TCI America

D1396-250G D1396-25G
TCD1396-250GEA 1286.12 CAD
TCD1396-250G TCD1396-025G
2,7-Dinitro-9-fluorenone ≥98.0%
2,7-Dinitro-9-fluorenone

CAS Number: 31551-45-8
MDL Number: MFCD00001153
Molecular Formula: C13H6N2O5
Molecular Weight: 270.20
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 298

Formula: C₁₃H₆N₂O₅
MW: 270.2 g/mol
Melting Pt: 296
MDL Number: MFCD00001153
CAS Number: 31551-45-8
EINECS: 250-695-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c0cf4ae410b52856750bcf21330a658997ed572&vid=5c0cf4ae410b52856750bcf21330a658997ed572&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR