1,2-Dimethylindole ≥98.0%

Supplier: TCI America

Synonyms: 1,2-Dimethyl-1H-indole, N,2-Dimethylindole

D1391-25G D1391-5G
TCD1391-25GEA 186.44 CAD
TCD1391-25G TCD1391-5G
1,2-Dimethylindole ≥98.0%
1,2-Dimethylindole

CAS Number: 875-79-6
MDL Number: MFCD00005802
Molecular Formula: C10H11N
Molecular Weight: 145.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 143
Melting point (°C): 55
Flash Point (°C): 110

Formula: C₁₀H₁₁N
Boiling Pt: 143 °C
Melting Pt: 55 °C
Flash Pt: 110 °C
MDL Number: MFCD00005802
CAS Number: 875-79-6
EINECS: 212-877-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c7ff5f89df464786a41f39b4045cf47abcf410b&vid=1c7ff5f89df464786a41f39b4045cf47abcf410b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR