2,2'-Dipyridylamine ≥99.0% (by GC, titration analysis)
Supplier: TCI America
D1200-5G
D1200-25G
TCD1200-5GEA
126.4
CAD
TCD1200-5G
TCD1200-025G
2,2'-Dipyridylamine ≥99.0% (by GC, titration analysis)
2,2'-Dipyridylamine
CAS Number: 1202-34-2
MDL Number: MFCD00006247
Molecular Formula: C10H9N3
Molecular Weight: 171.20
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 94
Flash Point (°C): 170
Other Product Names / Synonyms: 2,2'-Iminodipyridine
Formula:
C₁₀H₉N₃ Boiling Pt: 178 °C Melting Pt: 94 °C Flash Pt: 170 °C |
MDL Number:
MFCD00006247 CAS Number: 1202-34-2 EINECS: 214-864-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...