1,2-Diphenylethylamine ≥97.0%

Supplier: TCI America

Synonyms: alpha-Benzylbenzylamine, 1-Amino-1,2-diphenylethane

D1144-25G D1144-5G
TCD1144-25GEA 946.42 CAD
TCD1144-25G TCD1144-005G
1,2-Diphenylethylamine ≥97.0%
1,2-Diphenylethylamine

CAS Number: 25611-78-3
MDL Number: MFCD00008063
Molecular Formula: C14H15N
Molecular Weight: 197.28
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 1-Amino-1,2-diphenylethane, alpha-Benzylbenzylamine

Formula: C₁₄H₁₅N
MW: 197.28 g/mol
Density: 1.04 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008063
CAS Number: 25611-78-3
EINECS: 247-126-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef687b18e9e2a19146e6316d6584f21671833c48&vid=ef687b18e9e2a19146e6316d6584f21671833c48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR