2-(2,4-Dinitrobenzyl)pyridine ≥98.0%

Supplier: TCI America

D1095-5G D1095-1G
TCD1095-5GEA 211.56 CAD
TCD1095-5G TCD1095-001G
2-(2,4-Dinitrobenzyl)pyridine ≥98.0%
2-(2,4-Dinitrobenzyl)pyridine

CAS Number: 1151-97-9
MDL Number: MFCD00014632
Molecular Formula: C12H9N3O4
Molecular Weight: 259.22
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 90

Formula: C₁₂H₉N₃O₄
Melting Pt: 90 °C
MDL Number: MFCD00014632
CAS Number: 1151-97-9
EINECS: 214-567-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60d39dc4dc20b79d8cf615bfd3049f683c4c993a&vid=60d39dc4dc20b79d8cf615bfd3049f683c4c993a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR