1,4-Dimethylnaphthalene ≥96.0%
Supplier: TCI America
D1006-5G
D1006-25G
TCD1006-005GEA
221.78
CAD
TCD1006-005G
TCD1006-25G
1,4-Dimethylnaphthalene ≥96.0%
1,4-Dimethylnaphthalene
CAS Number: 571-58-4
MDL Number: MFCD00004037
Molecular Formula: C12H12
Molecular Weight: 156.23
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 268
Flash Point (°C): 128
Specific Gravity (20/20): 1.02
Formula:
C₁₂H₁₂ Boiling Pt: 268 °C Density: 1.02 (20 °C) Flash Pt: 128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004037 CAS Number: 571-58-4 EINECS: 209-335-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...