trans,trans-1,4-Diphenyl-1,3-butadiene ≥99.0%

Supplier: TCI America

Synonyms: (1E,3E)-1,4-Diphenylbuta-1,3-diene

D0879-5G D0879-25G
TCD0879-005GEA 85.54 CAD
TCD0879-005G TCD0879-025G
trans,trans-1,4-Diphenyl-1,3-butadiene ≥99.0%
trans,trans-1,4-Diphenyl-1,3-butadiene

CAS Number: 886-65-7
MDL Number: MFCD00004791
Molecular Formula: C16H14
Molecular Weight: 206.29
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 350
Melting point (°C): 152

Formula: C₁₆H₁₄
MW: 206.29 g/mol
Boiling Pt: 350 °C
Melting Pt: 152 °C
MDL Number: MFCD00004791
CAS Number: 538-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c303484b29c81e29388bacd356d89c5f75a1acd&vid=9c303484b29c81e29388bacd356d89c5f75a1acd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR