2-Methyl-2-propyl-1,3-propanediol ≥98.0%
Supplier: TCI America
Synonyms:
2,2-Bis(hydroxymethyl)pentane, 2-Methyl-2-propylpropane-1,3-diol, 2,2-Dimethylolpentane
D0766-25G
D0766-500G
TCD0766-25GEA
37.92
CAD
TCD0766-25G
TCD0766-500G
2-Methyl-2-propyl-1,3-propanediol ≥98.0%
2-Methyl-2-propyl-1,3-propanediol
CAS Number: 78-26-2
MDL Number: MFCD00004696
Molecular Formula: C7H16O2
Molecular Weight: 132.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 230
Melting point (°C): 57
Flash Point (°C): 130
Other Product Names / Synonyms: 2,2-Bis(hydroxymethyl)pentane, 2,2-Dimethylolpentane
Formula:
C₇H₁₆O₂ MW: 132.2 g/mol Boiling Pt: 230 °C Melting Pt: 57 °C Flash Pt: 130 °C |
MDL Number:
MFCD00004696 CAS Number: 78-26-2 EINECS: 201-099-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...