2,6-Dimethyl-4-heptanol ≥85.0%

Supplier: TCI America

Synonyms: Diisobutylcarbinol

D0732-25ML D0732-500ML
TCD0732-025MLEA 41.79 CAD
TCD0732-025ML TCD0732-500ML
2,6-Dimethyl-4-heptanol ≥85.0%
2,6-Dimethyl-4-heptanol

CAS Number: 108-82-7
MDL Number: MFCD00008944
Molecular Formula: C9H20O
Molecular Weight: 144.26
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 179
Flash Point (°C): 74
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: Diisobutylcarbinol

Formula: C₉H₂₀O
Boiling Pt: 179 °C
Melting Pt: –65 °C
Density: 0.81 (20 °C)
Flash Pt: 74 °C
MDL Number: MFCD00008944
CAS Number: 108-82-7
EINECS: 203-619-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f99e453f608d8577a43e3891a98a868baf042f06&vid=f99e453f608d8577a43e3891a98a868baf042f06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR