7,12-Dimethylbenz[a]anthracene ≥98.0%
Supplier: TCI America
D0677-1G
D0677-5G
TCD0677-001GEA
298.94
CAD
TCD0677-001G
TCD0677-5G
7,12-Dimethylbenz[a]anthracene ≥98.0%
7,12-Dimethylbenz[a]anthracene
CAS Number: 57-97-6
MDL Number: MFCD00003600
Molecular Formula: C20H16
Molecular Weight: 256.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 124
Formula:
C₂₀H₁₆ Boiling Pt: 440 °C (1013 hPa) Melting Pt: 124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003600 CAS Number: 57-97-6 EINECS: 200-359-5 UN: 3077 ADR: 9,III Merck Index: 12,03285 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...