Gramine ≥98.0%

Supplier: TCI America

Synonyms: 1-(1H-Indol-3-yl)-N,N-dimethylmethanamine, 3-(Dimethylaminomethyl)-Indole

D0653-25G D0653-250G
TCD0653-025GEA 54.64 CAD
TCD0653-025G TCD0653-250G
Gramine ≥98.0%
Gramine

CAS Number: 87-52-5
MDL Number: MFCD00005629
Molecular Formula: C11H14N2
Molecular Weight: 174.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 132
Flash Point (°C): 167


Product Categories: Bioscience, Alkaloids (Phytochemicals), Indole Alkaloids
Other Product Names / Synonyms: 3-(Dimethylaminomethyl)indole, Donaxine

Formula: C₁₁H₁₄N₂
Melting Pt: 132 °C
Flash Pt: 167 °C
MDL Number: MFCD00005629
CAS Number: 87-52-5
EINECS: 201-749-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ec9153996aef6674237ca86bcc0360d96abd356&vid=8ec9153996aef6674237ca86bcc0360d96abd356&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR