2,6-Dimethoxyphenol ≥98.0%
Supplier: TCI America
|
Warning
|
D0639-500G
D0639-25G
D0639-100G
TCD0639-500GEA
1896
CAD
TCD0639-500G
TCD0639-025G
TCD0639-100G
2,6-Dimethoxyphenol ≥98.0%
2,6-Dimethoxyphenol
CAS Number: 91-10-1
MDL Number: MFCD00064434
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 263
Melting point (°C): 56
Flash Point (°C): 78
Other Product Names / Synonyms: Pyrogallol 1,3-Dimethyl Ether
Formula:
C₈H₁₀O₃ MW: 154.17 g/mol Boiling Pt: 263 °C Melting Pt: 56 °C Flash Pt: 78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064434 CAS Number: 91-10-1 EINECS: 202-041-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...