1,4-Dihydroxynaphthalene ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D0589-5G D0589-25G
TCD0589-005GEA 183.44 CAD
TCD0589-005G TCD0589-025G
1,4-Dihydroxynaphthalene ≥97.0% (by GC, titration analysis)
1,4-Dihydroxynaphthalene

CAS Number: 571-60-8
MDL Number: MFCD00003977
Molecular Formula: C10H8O2
Molecular Weight: 160.17
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 190


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Naphthalenediol, alpha-Naphthohydroquinone

Formula: C₁₀H₈O₂
Melting Pt: 190 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003977
CAS Number: 571-60-8
EINECS: 209-336-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27fa6a6697ac4ccf793a43528f23eb15bbb3bed2&vid=27fa6a6697ac4ccf793a43528f23eb15bbb3bed2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR