Phenylhydroquinone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0584-5G D0584-25G
TCD0584-005GEA 576.86 CAD
TCD0584-005G TCD0584-025G
Phenylhydroquinone ≥98.0% (by GC, titration analysis)
Phenylhydroquinone

CAS Number: 1079-21-6
MDL Number: MFCD00002342
Molecular Formula: C12H10O2
Molecular Weight: 186.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 101


Other Product Names / Synonyms: Biphenyl-2,5-diol, 2,5-Dihydroxybiphenyl

Formula: C₁₂H₁₀O₂
Melting Pt: 101 °C
MDL Number: MFCD00002342
CAS Number: 1079-21-6
EINECS: 214-091-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=045e5395e4e0a6c499ccf2ce7db31534db18019c&vid=045e5395e4e0a6c499ccf2ce7db31534db18019c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR