3,5-Dihydroxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0570-25G D0570-100G
TCD0570-025GEA 66.36 CAD
TCD0570-025G TCD0570-100G
3,5-Dihydroxybenzoic acid ≥98.0% (by GC, titration analysis)
3,5-Dihydroxybenzoic acid

CAS Number: 99-10-5
MDL Number: MFCD00002512
Molecular Formula: C7H6O4
Molecular Weight: 154.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 200


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: alpha-Resorcylic Acid

Formula: C₇H₆O₄
Melting Pt: 236…238 °C
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002512
CAS Number: 99-10-5
EINECS: 202-730-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d6a2b2744e24796c4c804988b2f9e80b9b8c2f5&vid=5cce46643982ffbd621f8353d7a0cf73793477ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR