2,4-Dihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D0564-500G D0564-25G D0564-100G
TCD0564-500GEA 1483.62 CAD
TCD0564-500G TCD0564-025G TCD0564-100G
2,4-Dihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)
2,4-Dihydroxybenzaldehyde

CAS Number: 95-01-2
MDL Number: MFCD00011686
Molecular Formula: C7H6O3
Molecular Weight: 138.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 228
Melting point (°C): 137


Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction)
Other Product Names / Synonyms: beta-Resorcylaldehyde

Formula: C₇H₆O₃
Boiling Pt: 228 °C
Melting Pt: 137 °C
Density: 1.448 g/cm³ (–93 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011686
CAS Number: 95-01-2
EINECS: 202-383-1
Merck Index: 12,08324
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7083b8048e2c2f8e2e5dbcac079a80dd7d1ec8fa&vid=7850e7cf39d9197c204bdfea972cc5628b67e01d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR