9,10-Dihydrophenanthrene ≥96.0%
Supplier: TCI America
D0554-5G
D0554-25G
TCD0554-005GEA
205.4
CAD
TCD0554-005G
TCD0554-025G
9,10-Dihydrophenanthrene ≥96.0%
9,10-Dihydrophenanthrene
CAS Number: 776-35-2
MDL Number: MFCD00001164
Molecular Formula: C14H12
Molecular Weight: 180.25
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 314
Flash Point (°C): 110
Freezing point (°C): 32
Formula:
C₁₄H₁₂ Boiling Pt: 314 °C Melting Pt: 34 °C Density: 1.0953 g/cm³ (14 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001164 CAS Number: 776-35-2 EINECS: 212-278-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...