N,N-Diethyl-m-toluidine ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Diethylaminotoluene, N,N-Diethyl-3-methylaniline, DMT

D0532-500ML D0532-25ML
TCD0532-500MLEA 221.52 CAD
TCD0532-500ML TCD0532-025ML
N,N-Diethyl-m-toluidine ≥99.0% (by GC, titration analysis)
N,N-Diethyl-m-toluidine

CAS Number: 91-67-8
MDL Number: MFCD00035795
Molecular Formula: C11H17N
Molecular Weight: 163.26
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 107
Flash Point (°C): 104
Specific Gravity (20/20): 0.93


Other Product Names / Synonyms: 3-Diethylaminotoluene, N,N-Diethyl-3-methylaniline, DMT

Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Boiling Pt: 107 °C
Density: 0.93 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00035795
CAS Number: 91-67-8
EINECS: 202-089-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04b046e6d811ffbcbd810fcde497659b84712cc9&vid=04b046e6d811ffbcbd810fcde497659b84712cc9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR