2,6-Diethylaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
D0476-25ML
D0476-100ML
TCD0476-025MLEA
36.34
CAD
TCD0476-025ML
TCD0476-100ML
2,6-Diethylaniline ≥98.0% (by GC, titration analysis)
2,6-Diethylaniline
CAS Number: 579-66-8
MDL Number: MFCD00007753
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 216
Flash Point (°C): 115
Specific Gravity (20/20): 0.96
Formula:
C₁₀H₁₅N MW: 149.24 g/mol Boiling Pt: 216 °C Melting Pt: 3.5 Density: 0.96 (20 °C) Flash Pt: 115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007753 CAS Number: 579-66-8 EINECS: 209-445-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...