4,4'-Dithiodianiline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Bis(4-aminophenyl) disulphide, Di(4-aminophenyl) Disulfide, 4-Aminophenyl disulfide, 4-Aminophenyl Disulphide, Di(4-aminophenyl) Disulphide, 4-Aminophenyl disulphide, Bis(4-aminophenyl) disulfide, 4-Aminophenyl Disulfide, 4,4'-Diaminodiphenyl Disulfide, 4,4'-Diaminodiphenyl Disulphide
CAS Number: 722-27-0
MDL Number: MFCD00007882
Molecular Formula: C12H12N2S2
Molecular Weight: 248.36
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 75
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4-Aminophenyl Disulfide, 4,4'-Diaminodiphenyl Disulfide, Di(4-aminophenyl) Disulfide
Formula:
C₁₂H₁₂N₂S₂ MW: 248.37 g/mol Melting Pt: 75 °C |
MDL Number:
MFCD00007882 CAS Number: 722-27-0 EINECS: 211-961-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...